Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Quin-C7 | 871100-12-8 | 99.8% | 431.48 | C25H25N3O4 | 1 ML
SDP

Catalog No. 5000284824
Encompass_Preferred

Quin-C7 is an orally active antagonist of the formyl peptide receptor FPR2/ALX with reported anti-inflammatory activity in preclinical models. It has been used to ameliorate DSS-induced colitis in mice and is supplied for research use as either a 10 mM solution in DMSO (1 mL) or as a solid powder.

  • Orally active FPR2/ALX antagonist with anti-inflammatory activity.
  • Demonstrated efficacy in DSS-induced colitis mouse models.
  • Available as a 10 mM solution in DMSO (1 mL) or as a solid form.
  • High purity solid form (99.76%).
  • Molecular formula C25H25N3O4; molecular weight 431.48; CAS 871100-12-8.
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); solution at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-84824 Supplier Medchemexpress LLC Supplier No. HYW66877510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quin-C7 is an orally active antagonist of the formyl peptide receptor FPR2/ALX with reported anti-inflammatory activity in preclinical models. It has been used to ameliorate DSS-induced colitis in mice and is supplied for research use as either a 10 mM solution in DMSO (1 mL) or as a solid powder.

  • Orally active FPR2/ALX antagonist with anti-inflammatory activity.
  • Demonstrated efficacy in DSS-induced colitis mouse models.
  • Available as a 10 mM solution in DMSO (1 mL) or as a solid form.
  • High purity solid form (99.76%).
  • Molecular formula C25H25N3O4; molecular weight 431.48; CAS 871100-12-8.
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); solution at -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.