Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Quin-C7 | 871100-12-8 | MFCD34182190 | 99.8% | 431.48 g/mol | C25H25N3O4 | 10 MG
SDP

Numéro de catalogue. 5000284645
Encompass_Preferred

Quin-C7 is an orally active antagonist of formyl peptide receptor 2 (FPR2/ALX) used as a research reagent to study inflammatory signaling pathways. It exhibits anti-inflammatory activity and has been reported to ameliorate dextran sulfate sodium (DSS)-induced colitis in mouse models.

  • Orally active FPR2/ALX antagonist for in vivo and in vitro studies.
  • Demonstrated anti-inflammatory effects and efficacy in DSS-induced colitis models.
  • Chemical formula C25H25N3O4, molecular weight 431.48 g/mol.
  • CAS number 871100-12-8.
  • High purity (~99.8%) suitable for research applications.
  • Available as a 10 MG solid; soluble in DMSO for stock solutions.
  • For research use only; not for human or clinical use.

Numéro de catalogue. 50-002-84645 Fournisseur Medchemexpress LLC Code fournisseur HYW66877510MG
May include imposed supplier surcharges.
Ajouter au panier
Ajouter au panier

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Quin-C7 is an orally active antagonist of formyl peptide receptor 2 (FPR2/ALX) used as a research reagent to study inflammatory signaling pathways. It exhibits anti-inflammatory activity and has been reported to ameliorate dextran sulfate sodium (DSS)-induced colitis in mouse models.

  • Orally active FPR2/ALX antagonist for in vivo and in vitro studies.
  • Demonstrated anti-inflammatory effects and efficacy in DSS-induced colitis models.
  • Chemical formula C25H25N3O4, molecular weight 431.48 g/mol.
  • CAS number 871100-12-8.
  • High purity (~99.8%) suitable for research applications.
  • Available as a 10 MG solid; soluble in DMSO for stock solutions.
  • For research use only; not for human or clinical use.

missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.