Learn More
Medchemexpress LLC Quin-C7 | 871100-12-8 | MFCD34182190 | 99.8% | 431.48 g/mol | C25H25N3O4 | 100 MG
SDP
Catalog No. 5000284644
Encompass_Preferred
Quin-C7 is an orally active FPR2/ALX antagonist used in anti-inflammatory research, including dextran sulfate sodium (DSS)-induced colitis models. It is provided as a high-purity solid with characterized solubility and storage recommendations for both in vitro and in vivo studies.
  • Orally active FPR2/ALX antagonist for anti-inflammatory research.
  • Demonstrated efficacy in DSS-induced colitis models.
  • High purity (≈99.8%) for reliable experimental results.
  • White to off-white solid form, suitable for standard handling and storage.
  • Soluble in DMSO at high concentration and formulatable for in vivo dosing.
Catalog No. 50-002-84644 Supplier Medchemexpress LLC Supplier No. HYW668775100MG
Quin-C7 is an orally active FPR2/ALX antagonist used in anti-inflammatory research, including dextran sulfate sodium (DSS)-induced colitis models. It is provided as a high-purity solid with characterized solubility and storage recommendations for both in vitro and in vivo studies.
  • Orally active FPR2/ALX antagonist for anti-inflammatory research.
  • Demonstrated efficacy in DSS-induced colitis models.
  • High purity (≈99.8%) for reliable experimental results.
  • White to off-white solid form, suitable for standard handling and storage.
  • Soluble in DMSO at high concentration and formulatable for in vivo dosing.