Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Quin-C7 | 871100-12-8 | MFCD34182190 | 99.8% | 431.48 g/mol | C25H25N3O4 | 5 MG
SDP

Catalog No. 5000284037
Encompass_Preferred

Quin-C7 is an orally active antagonist of the formyl peptide receptor 2 (FPR2/ALX) with reported anti-inflammatory activity; it has been shown to ameliorate dextran sulfate sodium (DSS)-induced colitis in mice and is used in preclinical studies of inflammatory responses and inflammatory bowel disease.

  • Orally active FPR2/ALX antagonist used in inflammation research.
  • Validated in DSS-induced colitis models in mice.
  • High purity suitable for in vivo and in vitro studies (≈99.8%).
  • Molecular weight 431.48 g/mol; formula C25H25N3O4.
  • Supplied as a powder with COA and SDS available.
  • Recommended storage at -20°C to maintain stability.

Catalog No. 50-002-84037 Supplier Medchemexpress LLC Supplier No. HYW6687755MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quin-C7 is an orally active antagonist of the formyl peptide receptor 2 (FPR2/ALX) with reported anti-inflammatory activity; it has been shown to ameliorate dextran sulfate sodium (DSS)-induced colitis in mice and is used in preclinical studies of inflammatory responses and inflammatory bowel disease.

  • Orally active FPR2/ALX antagonist used in inflammation research.
  • Validated in DSS-induced colitis models in mice.
  • High purity suitable for in vivo and in vitro studies (≈99.8%).
  • Molecular weight 431.48 g/mol; formula C25H25N3O4.
  • Supplied as a powder with COA and SDS available.
  • Recommended storage at -20°C to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.