Learn More
Cayman Chemical QuInacrIn mustardhydroCLO 10mg
SDP
Catalog No. 502990519
Encompass_Preferred
A fluorescent DNA-intercalating agent; selectively binds to AT base pairs over GC base pairs; has been used to label metaphase chromosomes for karyotyping by autoradiography
Catalog No. 50-299-0519 Supplier Cayman Chemical Supplier No. 2965310 MG
A fluorescent DNA-intercalating agent; selectively binds to AT base pairs over GC base pairs; has been used to label metaphase chromosomes for karyotyping by autoradiography