Learn More
Cayman Chemical Quinacrine analog 34, Purity: ≥98%, CAS: 1411646-44-0, 10 mg
SDP
Catalog No. 502967411
Encompass_Preferred
An autophagy inhibitor (MEC = 0.5 µM in U2OS cells); induces lysosomal deacidification and accumulation of LC3-II in U2OS cells at 0.5 or 3 µM
Catalog No. 50-296-7411 Supplier Cayman Chemical Supplier No. 1773310 MG
An autophagy inhibitor (MEC = 0.5 µM in U2OS cells); induces lysosomal deacidification and accumulation of LC3-II in U2OS cells at 0.5 or 3 µM