Learn More
Apexbio Technology LLC Quinacrine (hydrochloride hydrate),100mg
SDP
Catalog No. 502240467
Encompass_Preferred
Quinacrine, also commonly known as atebrine, is a compound which is commonly used as an anti-protozoal agent. It inhibits voltage-dependent sodium channels with an IC50 value of 3.3 μM and suppresses aldehyde oxidase with an IC50 value of 3.3 μM. Quinacrine prevents misfolding of prion protein with an EC50 value of 0.3 μM. As an effective riboflavin antagonist, quinacrine associates with the riboflavin-binding protein with a Ki value of 6.7 μM. Voltage-dependent sodium channels play a vital role in action potential initiation and propagation in excitable cells, including muscle, nerve, and neuroendocrine cell types. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0467 Supplier Apexbio Technology LLC Supplier No. C5039100
Quinacrine, also commonly known as atebrine, is a compound which is commonly used as an anti-protozoal agent. It inhibits voltage-dependent sodium channels with an IC50 value of 3.3 μM and suppresses aldehyde oxidase with an IC50 value of 3.3 μM. Quinacrine prevents misfolding of prion protein with an EC50 value of 0.3 μM. As an effective riboflavin antagonist, quinacrine associates with the riboflavin-binding protein with a Ki value of 6.7 μM. Voltage-dependent sodium channels play a vital role in action potential initiation and propagation in excitable cells, including muscle, nerve, and neuroendocrine cell types. Other sizes are also available. Please inqury us for quote.