Learn More
Cayman Chemical Quinacrine (hydrochloride hydrate), 100 mg
SDP
Supplier:  Cayman Chemical 15041100
Encompass_Preferred
Molecular Formula C23H30ClN3O • 2HCl [XH2O], Purity ≥95%, Formula Weight 472.9, Synonyms: Atebrine, Mepacrine; An acridine compound with diverse biological activities and cell labeling properties; has anti-protozoal properties; prevents misfolding of prion protein (EC50 = 0.3 M); blocks voltage-dependent sodium channels (IC50 = 3.3 M); inhibits aldehyde oxidase (IC50 = 3.3 M); antagonizes the riboflavin-binding protein (Ki = 6.7 M); inhibits P-glycoprotein (EC50 = 14.4 M); stains DNA and RNA in fixed cells with ex/em = 436/525 nm, respectively; labels acidic vesicles, such as lysosomes, in live cells for long-term imaging with ex/em = 458/470-580 nm, respectively.
Catalog No. NC0664889