Learn More
Selleck Chemical LLC Quinacrine Dihydrochloride Dihydrate S5435-25mg
SDP
Catalog No. 503109780
Encompass_Preferred
Quinacrine (Mepacrine) is a dye of the acridine family that has been widely used as staining agents for DNA and model compounds for intercalators in numerous biophysical studies It is also an antimalarial drug
Catalog No. 50-310-9780 Supplier Selleck Chemical LLC Supplier No. S543525MG
Quinacrine (Mepacrine) is a dye of the acridine family that has been widely used as staining agents for DNA and model compounds for intercalators in numerous biophysical studies It is also an antimalarial drug