Learn More
Apexbio Technology LLC Quinacrine 2HCl 69-05-6 10mM (in 1mL H2O)
SDP
Catalog No. 5000092263
Encompass_Preferred
Quinacrine 2HCl (69-05-6) is an orally active small-molecule inhibitor targeting the NF- B signaling pathway It is designed to inhibit the NF- B cascade thereby downregulating pro-survival gene expression and promoting apoptotic pathways in tumor cells Quinacrine 2HCl exerts its biological activity primarily through inhibition of NF- B and activation of the p53-dependent apoptotic pathway In in vitro studies Quinacrine 2HCl demonstrates growth inhibition of multiple cancer cell lines with reported IC50 values generally in the low micromolar range depending on the specific cell model and experimental context Based on these pharmacological properties Quinacrine 2HCl holds research potential in investigating apoptosis regulatory mechanisms NF- B pathway involvement in cancer biology and preclinical evaluation of potential anticancer therapies
Catalog No. 50-000-92263 Supplier Apexbio Technology LLC Supplier No. B18235.1
Quinacrine 2HCl (69-05-6) is an orally active small-molecule inhibitor targeting the NF- B signaling pathway It is designed to inhibit the NF- B cascade thereby downregulating pro-survival gene expression and promoting apoptotic pathways in tumor cells Quinacrine 2HCl exerts its biological activity primarily through inhibition of NF- B and activation of the p53-dependent apoptotic pathway In in vitro studies Quinacrine 2HCl demonstrates growth inhibition of multiple cancer cell lines with reported IC50 values generally in the low micromolar range depending on the specific cell model and experimental context Based on these pharmacological properties Quinacrine 2HCl holds research potential in investigating apoptosis regulatory mechanisms NF- B pathway involvement in cancer biology and preclinical evaluation of potential anticancer therapies