Learn More
Medchemexpress LLC HY-13735A 100mg , Quinacrine (dihydrochloride) CAS:69-05-6 Purity:>98%
SDP
Catalog No. 502260094
Encompass_Preferred
HY-13735A 100mg Quinacrine (dihydrochloride) CAS: 69-05-6 Quinacrine (Mepacrine) dihydrochloride is an orally bioavailable antimalarial agent, which possess anticancer effect both in vitro and vivo. Quinacrine dihydrochloride suppresses NF-κB and activate p53 signaling, which results in the induction of the apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0094 Supplier Medchemexpress LLC Supplier No. HY13735A100MG
HY-13735A 100mg Quinacrine (dihydrochloride) CAS: 69-05-6 Quinacrine (Mepacrine) dihydrochloride is an orally bioavailable antimalarial agent, which possess anticancer effect both in vitro and vivo. Quinacrine dihydrochloride suppresses NF-κB and activate p53 signaling, which results in the induction of the apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.