Learn More
AdipoGen Quinacrine mustard 2HCl
SDP
Supplier:  AdipoGen CDXQ0011M010
Encompass_Preferred
Chemical. CAS 4213-45-0. Formula C23H28Cl3N3O . 2HCl. MW 468.9 . 72.9. Synthetic. Fluorescent probe for labeling chromosomal DNA. Also used in measurement of sequence specificity with Taq DNA polymerase.
Catalog No. 50-256-0164