Learn More
Abcam Quinacrine mustard dihydrochloride, Fluorescent probe for labeling chromosomal DNA, 10MG
Catalog No. 502705254
Encompass_Preferred
MW 541.8 g/mol, Purity >95%. Water-soluble nitrogen mustard analog of quinacrine (ab120749). Fluorescent probe for labeling chromosomal DNA. Fluoresces by reaction with nucleic acids. Also used in measurement of sequence specificity with Taq DNA polymerase.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5254 Supplier Abcam Supplier No. AB14537510MG
MW 541.8 g/mol, Purity >95%. Water-soluble nitrogen mustard analog of quinacrine (ab120749). Fluorescent probe for labeling chromosomal DNA. Fluoresces by reaction with nucleic acids. Also used in measurement of sequence specificity with Taq DNA polymerase.
The product is subject to the following: Abcam Restricted Use Statement