Learn More
Abcam Quinacrine mustard dihydrochloride, Fluorescent probe for labeling chromosomal DNA, 100MG
Catalog No. 502705255
Encompass_Preferred
MW 541.8 g/mol, Purity >95%. Water-soluble nitrogen mustard analog of quinacrine (ab120749). Fluorescent probe for labeling chromosomal DNA. Fluoresces by reaction with nucleic acids. Also used in measurement of sequence specificity with Taq DNA polymerase.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5255 Supplier Abcam Supplier No. AB145375100MG
Add to Cart
Add to Cart
MW 541.8 g/mol, Purity >95%. Water-soluble nitrogen mustard analog of quinacrine (ab120749). Fluorescent probe for labeling chromosomal DNA. Fluoresces by reaction with nucleic acids. Also used in measurement of sequence specificity with Taq DNA polymerase.
The product is subject to the following: Abcam Restricted Use Statement