Learn More
Medchemexpress LLC Quinagolide hydrochloride | 94424-50-7 | MFCD00893553 | 99.7% | C20H34ClN3O3S | 10MG
SDP
Catalog No. 5000258973
Encompass_Preferred
Quinagolide hydrochloride is the hydrochloride salt of quinagolide, a non-ergot-derived selective dopamine D2 receptor agonist used in research on prolactin regulation and dopaminergic signaling. Supplied as a research-grade solid with high purity for reliable in vitro and in vivo pharmacology studies.
  • Selective dopamine D2 receptor agonist for prolactin research.
  • Hydrochloride salt improves aqueous solubility.
  • High purity (≈99.7%) for reproducible results.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Available in research-scale pack sizes for dose-ranging experiments.
Catalog No. 50-002-58973 Supplier Medchemexpress LLC Supplier No. HY13736A10MG
Quinagolide hydrochloride is the hydrochloride salt of quinagolide, a non-ergot-derived selective dopamine D2 receptor agonist used in research on prolactin regulation and dopaminergic signaling. Supplied as a research-grade solid with high purity for reliable in vitro and in vivo pharmacology studies.
  • Selective dopamine D2 receptor agonist for prolactin research.
  • Hydrochloride salt improves aqueous solubility.
  • High purity (≈99.7%) for reproducible results.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Available in research-scale pack sizes for dose-ranging experiments.