Learn More
Medchemexpress LLC Quinagolide hydrochloride (CV205-502 hydrochloride) | 94424-50-7 | 99.6% | 432.02 | 100 MG
SDP
Catalog No. 5000351496
Encompass_Preferred
Quinagolide hydrochloride is a selective and orally active dopamine D2 receptor agonist. It inhibits prolactin and down-regulates AKT levels and its phosphorylation, demonstrating antitumor effects and making it suitable for cancer research.
  • Selective and orally active dopamine D2 receptor agonist
  • Inhibits prolactin
  • Down-regulates AKT levels and its phosphorylation
  • Shows antitumor effects
  • Reduces non-ergot dopamine receptor 2 (DRD2) mRNA expression
  • Inhibits invasive properties and endothelial differentiation of endometrial mesenchymal stromal cells (E-MSCs)
  • Effectively inhibits tumor growth in animal models
Catalog No. 50-003-51496 Supplier Medchemexpress LLC Supplier No. HY13736A100MG
Quinagolide hydrochloride is a selective and orally active dopamine D2 receptor agonist. It inhibits prolactin and down-regulates AKT levels and its phosphorylation, demonstrating antitumor effects and making it suitable for cancer research.
  • Selective and orally active dopamine D2 receptor agonist
  • Inhibits prolactin
  • Down-regulates AKT levels and its phosphorylation
  • Shows antitumor effects
  • Reduces non-ergot dopamine receptor 2 (DRD2) mRNA expression
  • Inhibits invasive properties and endothelial differentiation of endometrial mesenchymal stromal cells (E-MSCs)
  • Effectively inhibits tumor growth in animal models