Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinaldine, ≥90% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019007 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9007 500 mL
11-101-8462 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9007 Supplier MilliporeSigma Supelco Supplier No. Q2125500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Bottle
CAS 91-63-4
Density 1.058 g/mL (at 25°C)
Flash Point 79°C
Physical Form Neat
Linear Formula C10H9 N
MDL Number MFCD00006756
Melting Point -9°C to -3°C
Boiling Point 105°C to 107°C (10 mmHg); 248°C
Synonym 2-Methylquinoline
Type Analytical Standard
Quantity 500 mL
Formula Weight 143.19
Percent Purity ≥90% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.