Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinaldine phosphate salt, MilliporeSigma™ Supelco™

Catalog No. 111012706 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2706 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-2706 Supplier MilliporeSigma Supelco Supplier No. Q262625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 118896-93-8
Density 1.06 g/mL
Physical Form Neat
Linear Formula C10H9 N · H3 PO4
MDL Number MFCD00079536
Synonym 2-Methylquinoline
Type Analytical Standard
Quantity 25 g
Formula Weight 241.18
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.