Learn More
Medchemexpress LLC Quinaldine red | 117-92-0 | 97.0% | 5 G
SDP
Catalog No. 5000409769
Encompass_Preferred
Quinaldine red is a fluorescent probe used for determining the melting temperature (Tm) of proteins. It is intended for research use only.
  • Acts as a fluorescent probe
  • Used for determining the melting temperature (Tm) of proteins
Catalog No. 50-004-09769 Supplier Medchemexpress LLC Supplier No. HYD02675G
Quinaldine red is a fluorescent probe used for determining the melting temperature (Tm) of proteins. It is intended for research use only.
  • Acts as a fluorescent probe
  • Used for determining the melting temperature (Tm) of proteins