Learn More
Cayman Chemical QuInaldopeptIn 2.5mg
SDP
Catalog No. 502940857
Encompass_Preferred
A quinomycin antibiotic; active against a variety of bacteria, including S. pyogenes, S. aureus, C. perfringens, S. faecalis, E. coli, and K. pneumoniae (MICs = 0.2, 0.4, 0.8, 1.6, 3.1, and 6.3 UG/ml, respectively); cytotoxic to B16/F10 and Moser cells (IC50s = 0.0008 and 0.04 UG/ml, respectively); increases survival in a P388 leukemia mouse model; binds to and intercalates into DNA (Kd = 32 nM)
Catalog No. 50-294-0857 Supplier Cayman Chemical Supplier No. 281672.5 MG
A quinomycin antibiotic; active against a variety of bacteria, including S. pyogenes, S. aureus, C. perfringens, S. faecalis, E. coli, and K. pneumoniae (MICs = 0.2, 0.4, 0.8, 1.6, 3.1, and 6.3 UG/ml, respectively); cytotoxic to B16/F10 and Moser cells (IC50s = 0.0008 and 0.04 UG/ml, respectively); increases survival in a P388 leukemia mouse model; binds to and intercalates into DNA (Kd = 32 nM)