Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Quinapril hydrochloride | 82586-55-8 | 98.3% | 1 ML
SDP

Catalog No. 5000407941
Encompass_Preferred

Quinapril hydrochloride (CI-906) is a proagent in the angiotensin-converting enzyme (ACE) inhibitor class. It is used in the treatment of hypertension and congestive heart failure. Quinapril is rapidly de-esterified to quinaprilat, its active diacid metabolite, which is a potent ACE inhibitor.

  • Potent angiotensin converting enzyme (ACE) inhibitor
  • Used for hypertension and congestive heart failure treatment
  • Rapidly de-esterified to active quinaprilat metabolite
  • Appears as a white to off-white solid
  • Store solid form at 4°C, sealed and away from moisture
  • Store in solvent at -80°C for up to 6 months, or -20°C for 1 month, sealed and away from moisture
  • High solubility in DMSO (≥ 100 mg/mL) and H2O (≥ 50 mg/mL)

Catalog No. 50-004-07941 Supplier Medchemexpress LLC Supplier No. HYB047710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quinapril hydrochloride (CI-906) is a proagent in the angiotensin-converting enzyme (ACE) inhibitor class. It is used in the treatment of hypertension and congestive heart failure. Quinapril is rapidly de-esterified to quinaprilat, its active diacid metabolite, which is a potent ACE inhibitor.

  • Potent angiotensin converting enzyme (ACE) inhibitor
  • Used for hypertension and congestive heart failure treatment
  • Rapidly de-esterified to active quinaprilat metabolite
  • Appears as a white to off-white solid
  • Store solid form at 4°C, sealed and away from moisture
  • Store in solvent at -80°C for up to 6 months, or -20°C for 1 month, sealed and away from moisture
  • High solubility in DMSO (≥ 100 mg/mL) and H2O (≥ 50 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.