Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Quinapril (hydrochloride) | 82586-55-8 | 98.3% | 500 MG
SDP

Catalog No. 5000406383
Encompass_Preferred

Quinapril hydrochloride (CI-906) is a proagent belonging to the angiotensin-converting enzyme (ACE) inhibitor class of medications. It is used in the treatment of hypertension and congestive heart failure. Quinapril is rapidly de-esterified after absorption to quinaprilat (the active diacid metabolite), a potent ACE inhibitor.

  • Molecular weight: 474.98
  • Formula: C25H31ClN2O5
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture)
  • Solubility in DMSO: ≥ 100 mg/mL
  • Solubility in H2O: ≥ 50 mg/mL

Catalog No. 50-004-06383 Supplier Medchemexpress LLC Supplier No. HYB0477500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quinapril hydrochloride (CI-906) is a proagent belonging to the angiotensin-converting enzyme (ACE) inhibitor class of medications. It is used in the treatment of hypertension and congestive heart failure. Quinapril is rapidly de-esterified after absorption to quinaprilat (the active diacid metabolite), a potent ACE inhibitor.

  • Molecular weight: 474.98
  • Formula: C25H31ClN2O5
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture)
  • Solubility in DMSO: ≥ 100 mg/mL
  • Solubility in H2O: ≥ 50 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.