Learn More
Cayman Chemical Quinapril (hydrochloride), Purity: ≥98%, CAS: 82586-55-8, 500 mg
SDP
Catalog No. 502994286
Encompass_Preferred
A prodrug form of quinaprilat; reduces mean arterial pressure in renal hypertensive and spontaneously hypertensive rats at 3 mg/kg; inhibits angiotensin I-induced pressor responses in normotensive rats and dogs; prevents left ventricular heart failure in CHF 14.6 cardiomyopathic hamsters
Catalog No. 50-299-4286 Supplier Cayman Chemical Supplier No. 21439500 MG
A prodrug form of quinaprilat; reduces mean arterial pressure in renal hypertensive and spontaneously hypertensive rats at 3 mg/kg; inhibits angiotensin I-induced pressor responses in normotensive rats and dogs; prevents left ventricular heart failure in CHF 14.6 cardiomyopathic hamsters