Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Quinazolin-4-ylthiazol-2-ylamine | 404828-14-4 | 98.8% | 301.35 g/mol | C18H15N5 | 5 MG
SDP

Catalog No. 5000263274
Encompass_Preferred

GSK-3 Inhibitor XIII is a potent, ATP-competitive small-molecule inhibitor of glycogen synthase kinase-3 (GSK-3) with a reported Ki of 24 nM. Supplied as a high-purity solid, it is intended for biochemical kinase assays and cellular studies of GSK-3 signaling and preclinical research.

  • Potent ATP-competitive GSK-3 inhibitor (Ki 24 nM).
  • High purity: 98.79% as supplied.
  • Soluble in DMSO (100 mg/mL) and in formulation (10% DMSO/40% PEG300/5% Tween-80/45% saline) ≥ 2.5 mg/mL.
  • Solid appearance: white to off-white powder, molecular weight 301.35 g/mol.
  • Storage: powder -20°C (up to 3 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Demonstrated antiproliferative activity across multiple cell lines in cellular assays.

Catalog No. 50-002-63274 Supplier Medchemexpress LLC Supplier No. HY1123925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GSK-3 Inhibitor XIII is a potent, ATP-competitive small-molecule inhibitor of glycogen synthase kinase-3 (GSK-3) with a reported Ki of 24 nM. Supplied as a high-purity solid, it is intended for biochemical kinase assays and cellular studies of GSK-3 signaling and preclinical research.

  • Potent ATP-competitive GSK-3 inhibitor (Ki 24 nM).
  • High purity: 98.79% as supplied.
  • Soluble in DMSO (100 mg/mL) and in formulation (10% DMSO/40% PEG300/5% Tween-80/45% saline) ≥ 2.5 mg/mL.
  • Solid appearance: white to off-white powder, molecular weight 301.35 g/mol.
  • Storage: powder -20°C (up to 3 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Demonstrated antiproliferative activity across multiple cell lines in cellular assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.