Learn More
Medchemexpress LLC Quinazoline, 6,7-dimethoxy-4-[(3R)-3-(2-quinoxalinyloxy)-1-pyrrolidinyl]- | 927691-21-2 | 99.8% | 403.43 | 50 MG
SDP
Catalog No. 5000386990
Encompass_Preferred
PQ-10 is a potent inhibitor of Phosphodiesterase 10A (PDE10A) with IC50 and ED50 values of 4.6 nM and 13 mg/kg, respectively. It induces patterns of brain glucose metabolism, which can serve as a potential translational biomarker. It has the potential for researching psychiatric disorders like schizophrenia.
  • Potent inhibitor of Phosphodiesterase 10A (PDE10A)
  • IC50 of 4.6 nM
  • ED50 of 13 mg/kg
  • Induces patterns of brain glucose metabolism
  • Potential translational biomarker
  • Potential for researching psychiatric disorders like schizophrenia
Catalog No. 50-003-86990 Supplier Medchemexpress LLC Supplier No. HY1807850MG
PQ-10 is a potent inhibitor of Phosphodiesterase 10A (PDE10A) with IC50 and ED50 values of 4.6 nM and 13 mg/kg, respectively. It induces patterns of brain glucose metabolism, which can serve as a potential translational biomarker. It has the potential for researching psychiatric disorders like schizophrenia.
  • Potent inhibitor of Phosphodiesterase 10A (PDE10A)
  • IC50 of 4.6 nM
  • ED50 of 13 mg/kg
  • Induces patterns of brain glucose metabolism
  • Potential translational biomarker
  • Potential for researching psychiatric disorders like schizophrenia