Learn More
Medchemexpress LLC Quinelorane dihydrochloride | 97548-97-5 | MFCD00865952 | 99.0% | 319.27 g/mol | C14H24Cl2N4 | 5 MG
SDP
Catalog No. 5000245142
Encompass_Preferred
Quinelorane dihydrochloride (LY163502) is a selective dopamine D3/D2 receptor agonist supplied for preclinical neuropharmacology research. Provided as the dihydrochloride salt, it is a white to off-white solid (molecular weight 319.27 g/mol, CAS 97548-97-5) typically supplied at high purity for laboratory use.
  • Selective dopamine D3/D2 receptor agonist for pharmacological studies.
  • Supplied as the dihydrochloride salt in a stable solid form.
  • High purity (~99.0%) appropriate for research applications.
  • Storage: solid at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).
  • Commonly provided in small research quantities suitable for in vitro and in vivo experiments.
Catalog No. 50-002-45142 Supplier Medchemexpress LLC Supplier No. HY1034295MG
Quinelorane dihydrochloride (LY163502) is a selective dopamine D3/D2 receptor agonist supplied for preclinical neuropharmacology research. Provided as the dihydrochloride salt, it is a white to off-white solid (molecular weight 319.27 g/mol, CAS 97548-97-5) typically supplied at high purity for laboratory use.
  • Selective dopamine D3/D2 receptor agonist for pharmacological studies.
  • Supplied as the dihydrochloride salt in a stable solid form.
  • High purity (~99.0%) appropriate for research applications.
  • Storage: solid at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).
  • Commonly provided in small research quantities suitable for in vitro and in vivo experiments.