Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinestrol, TCI America™
SDP

Catalog No. Q0091100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q0091100MG 100 mg
Q00911G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q0091100MG Supplier TCI America Supplier No. Q0091100MG

Chemical Identifiers

CAS 152-43-2
Molecular Formula C25H32O2
Molecular Weight (g/mol) 364.53
MDL Number MFCD00079189
InChI Key PWZUUYSISTUNDW-VAFBSOEGSA-N
Synonym 17alpha-Ethynylestradiol 3-Cyclopentyl Ether
PubChem CID 9046
ChEBI CHEBI:8716
IUPAC Name (1R,3aS,3bR,9bS,11aS)-7-(cyclopentyloxy)-1-ethynyl-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-ol
SMILES C[C@]12CC[C@H]3[C@@H](CCC4=CC(OC5CCCC5)=CC=C34)[C@@H]1CC[C@@]2(O)C#C

Specifications

Melting Point 110°C
Color White
Quantity 100 mg
Formula Weight 364.53
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Quinestrol
RTECSNumber RC8948000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.