Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinhydrone, 98.37%, MP Biomedicals™

Catalog No. icn15648225 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN15648225 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15648225 Supplier MP Biomedicals, Inc Supplier No. 0215648225
Only null left
Add to Cart
Add to Cart

Specifications

CAS 106-34-3
Color Green
Assay Percent Range 0.9837
Molecular Formula C12H10O4
Quantity 25 g
MDL Number MFCD00010310
Synonym quinhydrone, green hydroquinone, p-benzoquinhydrone, chinhydron, chinhydron czech, unii-p4a66lq3qj, hydroquinone, compd. with p-benzoquinone, p-benzoquinone, compd. with hydroquinone, 2,5-cyclohexadiene-1,4-dione, compd. with 1,4-benzenediol 1:1, p-benzoquinone-hydroquinone compound 1:1
InChI Key BDJXVNRFAQSMAA-UHFFFAOYSA-N
SMILES OC1=CC=C(O)C=C1.O=C1C=CC(=O)C=C1
IUPAC Name benzene-1,4-diol; cyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol) 218.21
PubChem CID 7801
ChEBI CHEBI:26491
Formula Weight 218.2
Physical Form Crystalline Powder
Chemical Name or Material Quinhydrone
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.