Learn More
Sigma Aldrich Fine Chemicals Biosciences Fluorescein isothiocyanate–dextran average mol wt 4,000, (FITC:Glucose = 1:250) | 60842-46-8 | MFCD00131092 | 500mg
Catalog No. 501780499
Encompass
Fluorescein isothiocyanate–dextran average mol wt 4,000, (FITC:Glucose = 1:250)MW: | 60842-46-8 | MFCD00131092 | 500mg
Catalog No. 50-178-0499 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 46944500MGF
Fluorescein isothiocyanate–dextran average mol wt 4,000, (FITC:Glucose = 1:250)MW: | 60842-46-8 | MFCD00131092 | 500mg