Learn More
Medchemexpress LLC Quinidine (15% dihydroquinidine) | 56-54-2 | >98.36% | 10 G
SDP
Catalog No. 5000411674
Encompass_Preferred
Quinidine (15% dihydroquinidine) is an antiarrhythmic agent. It acts as a potent, orally active, and selective cytochrome P450db inhibitor. Additionally, Quinidine is a K+ channel blocker with an IC50 of 19.9 μM and is capable of inducing apoptosis. This compound can also be utilized for malaria research.
  • Antiarrhythmic agent.
  • Potent, orally active, selective cytochrome P450db inhibitor.
  • K+ channel blocker with an IC50 of 19.9 μM.
  • Can induce apoptosis.
  • Used for malaria research.
Catalog No. 50-004-11674 Supplier Medchemexpress LLC Supplier No. HYB175110G
Quinidine (15% dihydroquinidine) is an antiarrhythmic agent. It acts as a potent, orally active, and selective cytochrome P450db inhibitor. Additionally, Quinidine is a K+ channel blocker with an IC50 of 19.9 μM and is capable of inducing apoptosis. This compound can also be utilized for malaria research.
  • Antiarrhythmic agent.
  • Potent, orally active, selective cytochrome P450db inhibitor.
  • K+ channel blocker with an IC50 of 19.9 μM.
  • Can induce apoptosis.
  • Used for malaria research.