Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Quinidine (15% dihydroquinidine) | 56-54-2 | 98.36% | 5 G
SDP

Catalog No. 5000412860
Encompass_Preferred

Quinidine (15% dihydroquinidine) is an antiarrhythmic agent. It is a potent, orally active, selective cytochrome P450db inhibitor. Quinidine also acts as a K+ channel blocker with an IC50 of 19.9 μM, and it can induce apoptosis. It can be used for malaria research.

  • Antiarrhythmic agent: Helps in the treatment of irregular heartbeats.
  • Potent, orally active, selective cytochrome P450db inhibitor: Targets and inhibits specific cytochrome P450 enzymes.
  • K+ channel blocker: Blocks potassium channels with an IC50 of 19.9 μM.
  • Induces apoptosis: Capable of triggering programmed cell death.
  • Used for malaria research: Applicable in studies related to malaria.

Catalog No. 50-004-12860 Supplier Medchemexpress LLC Supplier No. HYB17515G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quinidine (15% dihydroquinidine) is an antiarrhythmic agent. It is a potent, orally active, selective cytochrome P450db inhibitor. Quinidine also acts as a K+ channel blocker with an IC50 of 19.9 μM, and it can induce apoptosis. It can be used for malaria research.

  • Antiarrhythmic agent: Helps in the treatment of irregular heartbeats.
  • Potent, orally active, selective cytochrome P450db inhibitor: Targets and inhibits specific cytochrome P450 enzymes.
  • K+ channel blocker: Blocks potassium channels with an IC50 of 19.9 μM.
  • Induces apoptosis: Capable of triggering programmed cell death.
  • Used for malaria research: Applicable in studies related to malaria.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.