Learn More
Medchemexpress LLC Quinidine (15% dihydroquinidine) | 56-54-2 | 98.36% | 500 MG
SDP
Catalog No. 5000412712
Encompass_Preferred
Quinidine (15% dihydroquinidine) is an antiarrhythmic agent. It acts as a potent, orally active, selective cytochrome P450db inhibitor and a K+ channel blocker. It induces apoptosis and can be utilized for malaria research. It has demonstrated cytotoxicity against MES-SA cells.
  • Antiarrhythmic agent
  • Potent, orally active, selective cytochrome P450db inhibitor
  • K+ channel blocker
  • Induces apoptosis
  • Can be used for malaria research
Catalog No. 50-004-12712 Supplier Medchemexpress LLC Supplier No. HYB1751500MG
Quinidine (15% dihydroquinidine) is an antiarrhythmic agent. It acts as a potent, orally active, selective cytochrome P450db inhibitor and a K+ channel blocker. It induces apoptosis and can be utilized for malaria research. It has demonstrated cytotoxicity against MES-SA cells.
  • Antiarrhythmic agent
  • Potent, orally active, selective cytochrome P450db inhibitor
  • K+ channel blocker
  • Induces apoptosis
  • Can be used for malaria research