Learn More
Medchemexpress LLC Quinidine (15% dihydroquinidine) | 56-54-2 | 98.4% | 25 G
SDP
Catalog No. 5000414269
Encompass_Preferred
Quinidine (15% dihydroquinidine) is an antiarrhythmic agent. It is a potent, orally active, selective cytochrome P450db inhibitor and a K+ channel blocker. It can induce apoptosis and is used for malaria research.
  • Antiarrhythmic agent
  • Potent, orally active, selective cytochrome P450db inhibitor
  • K+ channel blocker with an IC50 of 19.9 μM
  • Induces apoptosis
  • Can be used for malaria research
  • Shows cytotoxicity against MES-SA cells
  • Shows effects on the PTZ-induced seizure threshold in vivo
Catalog No. 50-004-14269 Supplier Medchemexpress LLC Supplier No. 25GHYB1751
Quinidine (15% dihydroquinidine) is an antiarrhythmic agent. It is a potent, orally active, selective cytochrome P450db inhibitor and a K+ channel blocker. It can induce apoptosis and is used for malaria research.
  • Antiarrhythmic agent
  • Potent, orally active, selective cytochrome P450db inhibitor
  • K+ channel blocker with an IC50 of 19.9 μM
  • Induces apoptosis
  • Can be used for malaria research
  • Shows cytotoxicity against MES-SA cells
  • Shows effects on the PTZ-induced seizure threshold in vivo