Learn More
AdipoGen Quinidine anhydrous
SDP
Supplier:  AdipoGen CDXQ0029G050
Encompass_Preferred
Chemical. CAS 56-54-2. Formula C20H24N2O2. MW BD9837. Quinidine is a dextrorotatory stereoisomer of Quinine with anti-malarial, anti-arrhythmic class 1A, anti-inflammatory, anti-cancer and anti-viral properties. Quinidine acts as a blocker of voltage-gated sodium channels. Inhibition of the Nav1.5 channel is specifically involved in its anti-arrhythmic effects as a class I anti-arrhythmic agent. Quinidine also blocks certain voltage-gated potassium channels e.g. Kv1.4, Kv4.2, hERG, among others, acts as an anti-muscarinic. Quinidine blocks alpha1-adrenoceptors alpha1ARs and is an inhibitor of cytochrome P450 and acetylcholinesterase.
Catalog No. 50-256-0154