Learn More
Selleck Chemical LLC Quinidine hydrochloride monohydrate S4681-25mg
SDP
Catalog No. 503109368
Encompass_Preferred
Quinidine hydrochloride monohydrate is an anti-arrythmic agent and a potent potassium channel inhibitor with IC50 of 19 9 M
Catalog No. 50-310-9368 Supplier Selleck Chemical LLC Supplier No. S468125MG
Quinidine hydrochloride monohydrate is an anti-arrythmic agent and a potent potassium channel inhibitor with IC50 of 19 9 M