Learn More
Selleck Chemical LLC Quinidine hydrochloride monohydrate S4681-1g
SDP
Catalog No. 503111456
Encompass_Preferred
Quinidine hydrochloride monohydrate is an anti-arrythmic agent and a potent potassium channel inhibitor with IC50 of 19 9 M
Catalog No. 50-311-1456 Supplier Selleck Chemical LLC Supplier No. S46811G
Quinidine hydrochloride monohydrate is an anti-arrythmic agent and a potent potassium channel inhibitor with IC50 of 19 9 M