Learn More
Medchemexpress LLC Quinidine hydrochloride monohydrate | 6151-40-2 | 99.97% | 100 MG
SDP
Catalog No. 5000412095
Encompass_Preferred
Quinidine hydrochloride monohydrate is an anti-arrhythmic agent that also acts as a potent blocker of K+ channels. It is intended for research use only.
  • Potent K+ channel blocker.
  • Aids in electrophysiological research.
  • Available in solid and solution forms for various experimental needs.
  • Comes with detailed documentation for quality assurance.
  • Referenced in numerous scientific publications.
  • Offers diverse applications for in vitro and in vivo studies.
  • Easily soluble in common laboratory solvents.
  • Supports accurate dosage calculations for experimental protocols.
Catalog No. 50-004-12095 Supplier Medchemexpress LLC Supplier No. HYB130210MM/1ML
Quinidine hydrochloride monohydrate is an anti-arrhythmic agent that also acts as a potent blocker of K+ channels. It is intended for research use only.
  • Potent K+ channel blocker.
  • Aids in electrophysiological research.
  • Available in solid and solution forms for various experimental needs.
  • Comes with detailed documentation for quality assurance.
  • Referenced in numerous scientific publications.
  • Offers diverse applications for in vitro and in vivo studies.
  • Easily soluble in common laboratory solvents.
  • Supports accurate dosage calculations for experimental protocols.