Learn More
Selleck Chemical LLC Quinidine sulfate 25mg 6591-63-5 Chinidin Sodium, Pitayine Sodium, -quinine Sodium, (+)-quinidine Sodium
SDP
Supplier:  Selleck Chemical LLC S375125MG
Encompass_Preferred
Quinidine (Chinidin, Pitayine, ?-quinine, (+)-quinidine) acts as a blocker of voltage-gated sodium channels, also an antimuscarinic and antimalarial. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8401