Learn More
Apexbio Technology LLC QUINIDINE-SULFATE-50MG
SDP
Catalog No. 5000568613
Encompass_Preferred
Quinidine sulfate (CAS No 6591-63-5) is a pharmaceutical compound primarily used as an antiarrhythmic agent in biomedical research Originating from the bark of the cinchona tree its mechanism of action involves blocking sodium and potassium channels thereby affecting cardiac action potentials This modulation helps prevent abnormal heart rhythms In vitro quinidine sulfate exhibits activity in the nanomolar to low micromolar range which indicates its potency in disrupting ionic channels within cells It is commonly used in various experimental applications including cell models animal studies and drug screening to assess cardiac and electrophysiological effects Research often employs this compound to explore new therapeutic strategies for arrhythmia and related cardiovascular conditions Experimental concentrations typically depend on the specific research design allowing flexibility based on the investigative focus and model system employed
Catalog No. 50-005-68613 Supplier Apexbio Technology LLC Supplier No. C646250
Quinidine sulfate (CAS No 6591-63-5) is a pharmaceutical compound primarily used as an antiarrhythmic agent in biomedical research Originating from the bark of the cinchona tree its mechanism of action involves blocking sodium and potassium channels thereby affecting cardiac action potentials This modulation helps prevent abnormal heart rhythms In vitro quinidine sulfate exhibits activity in the nanomolar to low micromolar range which indicates its potency in disrupting ionic channels within cells It is commonly used in various experimental applications including cell models animal studies and drug screening to assess cardiac and electrophysiological effects Research often employs this compound to explore new therapeutic strategies for arrhythmia and related cardiovascular conditions Experimental concentrations typically depend on the specific research design allowing flexibility based on the investigative focus and model system employed