Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quinidine Sulfate, Dihydrate, USP, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18606019 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-019 10 g Amber Glass Bottle
18-606-020 50 g Amber Glass Bottle
18-606-021 100 g Amber Glass Bottle
18-606-022 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-019 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. QU10510GM

QU105, 6591-63-5, (C20H24N2O2)2·H2SO4·2H2O

Spectrum™ Chemical Quinidine Sulfate, Dihydrate, USP is an antimalarial and antiarrhythmic drug used to treat or prevent many types of irregular heartbeats. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6591-63-5
Molecular Formula C40H54N4O10S
Molecular Weight (g/mol) 782.95
InChI Key ZHNFLHYOFXQIOW-YKSBFIEBSA-N
IUPAC Name bis((S)-[(2R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](6-methoxyquinolin-4-yl)methanol) sulfuric acid dihydrate
SMILES O.O.OS(O)(=O)=O.COC1=CC=C2N=CC=C([C@H](O)[C@H]3CC4CCN3C[C@@H]4C=C)C2=C1.COC1=CC=C2N=CC=C([C@H](O)[C@H]3CC4CCN3C[C@@H]4C=C)C2=C1

Specifications

CAS Max % 1
Ignition Residue 0.001
Packaging Amber Glass Bottle
Quantity 10 g
Specific Rotation +275° to +288°
Percent Purity 99 to 101%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.