Learn More
Crescent Chemical Co Inc QUININE 100 MG
SDP
Catalog No. 5076873
Encompass_Preferred
Quinine Pharma./veter. comp. (WS) CAS 130-95-0 Molecular Weight - 324.4168 Storage - RT Catalog Num - DRE-C16706900 chemical
Catalog No. 50-768-73 Supplier Crescent Chemical Co Inc Supplier No. PSC16706900
Quinine Pharma./veter. comp. (WS) CAS 130-95-0 Molecular Weight - 324.4168 Storage - RT Catalog Num - DRE-C16706900 chemical