Learn More
Selleck Chemical LLC Quinine S4495-25mg
SDP
Catalog No. 503112108
Encompass_Preferred
Quinine an alkaloid derived from the bark of the cinchona tree is an anti-malaria agent and a potassium channel inhibitor that inhibits mSlo3 (KCa 5 1) channel currents evoked by voltage pulses to 100 mV with IC50 of 169 M
Catalog No. 50-311-2108 Supplier Selleck Chemical LLC Supplier No. S449525MG
Quinine an alkaloid derived from the bark of the cinchona tree is an anti-malaria agent and a potassium channel inhibitor that inhibits mSlo3 (KCa 5 1) channel currents evoked by voltage pulses to 100 mV with IC50 of 169 M