Learn More
Medchemexpress LLC Quinine | 130-95-0 | 25 G
SDP
Catalog No. 5000414288
Encompass_Preferred
Quinine is an alkaloid derived from the bark of the cinchona tree, acting as an anti-malaria agent and a potassium channel inhibitor. It inhibits WT mouse Slo3 (KCa5.1) channel currents evoked by voltage pulses to +100 mV with an IC50 of 169 μM.
  • Derived from cinchona tree bark
  • Acts as an anti-malaria agent
  • Functions as a potassium channel inhibitor
  • Inhibits WT mouse Slo3 (KCa5.1) channel currents
Catalog No. 50-004-14288 Supplier Medchemexpress LLC Supplier No. 25GHYD0143
Quinine is an alkaloid derived from the bark of the cinchona tree, acting as an anti-malaria agent and a potassium channel inhibitor. It inhibits WT mouse Slo3 (KCa5.1) channel currents evoked by voltage pulses to +100 mV with an IC50 of 169 μM.
  • Derived from cinchona tree bark
  • Acts as an anti-malaria agent
  • Functions as a potassium channel inhibitor
  • Inhibits WT mouse Slo3 (KCa5.1) channel currents