Learn More
Medchemexpress LLC Quinine | 130-95-0 | 99.8% | 10 G
SDP
Catalog No. 5000410787
Encompass_Preferred
Quinine is an alkaloid derived from the bark of the cinchona tree, acting as an anti-malaria agent. It functions as a potassium channel inhibitor, specifically inhibiting WT mouse Slo3 (KCa5.1) channel currents evoked by voltage pulses to +100 mV with an IC50 of 169 μM. It is for research use only and not sold to patients.
  • Anti-malaria agent
  • Potassium channel inhibitor (inhibits WT mouse Slo3 (KCa5.1) channel currents with an IC50 of 169 μM)
  • Inhibits DENV virus replication in HepG2 cells
  • Reduces DENV-positive cells in a dose-dependent manner
  • Shows tumor-suppressing effects on skin cancer in Swiss albino mice
Catalog No. 50-004-10787 Supplier Medchemexpress LLC Supplier No. HYD014310G
Quinine is an alkaloid derived from the bark of the cinchona tree, acting as an anti-malaria agent. It functions as a potassium channel inhibitor, specifically inhibiting WT mouse Slo3 (KCa5.1) channel currents evoked by voltage pulses to +100 mV with an IC50 of 169 μM. It is for research use only and not sold to patients.
  • Anti-malaria agent
  • Potassium channel inhibitor (inhibits WT mouse Slo3 (KCa5.1) channel currents with an IC50 of 169 μM)
  • Inhibits DENV virus replication in HepG2 cells
  • Reduces DENV-positive cells in a dose-dependent manner
  • Shows tumor-suppressing effects on skin cancer in Swiss albino mice