Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Quinine | 130-95-0 | 99.8% | 5 G
SDP

Catalog No. 5000410519
Encompass_Preferred

Quinine is an alkaloid derived from the bark of the cinchona tree, functioning as an anti-malaria agent. It acts as a potassium channel inhibitor, specifically inhibiting WT mouse Slo3 (KCa5.1) channel currents with an IC50 of 169 μM. For optimal stability, store as a solid at 4°C, protected from light, and under nitrogen.

  • Acts as an anti-malaria agent
  • Functions as a potassium channel inhibitor
  • Purity of 99.82%
  • Molecular weight of 324.42
  • Chemical formula C20H24N2O2
  • Appears as a white to off-white solid

Catalog No. 50-004-10519 Supplier Medchemexpress LLC Supplier No. HYD01435G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Quinine is an alkaloid derived from the bark of the cinchona tree, functioning as an anti-malaria agent. It acts as a potassium channel inhibitor, specifically inhibiting WT mouse Slo3 (KCa5.1) channel currents with an IC50 of 169 μM. For optimal stability, store as a solid at 4°C, protected from light, and under nitrogen.

  • Acts as an anti-malaria agent
  • Functions as a potassium channel inhibitor
  • Purity of 99.82%
  • Molecular weight of 324.42
  • Chemical formula C20H24N2O2
  • Appears as a white to off-white solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.