Learn More
Medchemexpress LLC HY-D0143 1g Medchemexpress, Quinine CAS:130-95-0 Purity:>98%
SDP
Catalog No. 501933150
Encompass_Preferred
Medchemexpress, HY-D0143 1g Quinine CAS:130-95-0 Quinine is an anti-malaria agent and also a potassium channel inhibitor with an IC 50 of 169 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3150 Supplier Medchemexpress LLC Supplier No. HYD01431G
Medchemexpress, HY-D0143 1g Quinine CAS:130-95-0 Quinine is an anti-malaria agent and also a potassium channel inhibitor with an IC 50 of 169 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.