Learn More
AdipoGen Quinine anhydrous
SDP
Catalog No. 502560161
Encompass_Preferred
Chemical. CAS 130-95-0. Formula C20H24N2O2. MW 324.42. Synthetic. Because of its relatively constant and well-known fluorescence quantum yield, quinine is also used in photochemistry as a common fluorescence standard. It has been used for imaging of oxygen evolution and oxide formation. Chloride and bromide have been shown to quench fluorescence. Generally it is famous as potassium channel blocker with antipyretic fever-reducing, antimalarial, analgesic painkilling, and anti-inflammatory properties.
Catalog No. 50-256-0161 Supplier AdipoGen Supplier No. CDXQ0012G005
Chemical. CAS 130-95-0. Formula C20H24N2O2. MW 324.42. Synthetic. Because of its relatively constant and well-known fluorescence quantum yield, quinine is also used in photochemistry as a common fluorescence standard. It has been used for imaging of oxygen evolution and oxide formation. Chloride and bromide have been shown to quench fluorescence. Generally it is famous as potassium channel blocker with antipyretic fever-reducing, antimalarial, analgesic painkilling, and anti-inflammatory properties.