Learn More
TARGETMOL CHEMICALS INC Quinine dihydrochloride 500MG
SDP
Catalog No. 502878932
Encompass_Preferred
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Quinine dihydrochloride (Quinine bimuriate) is a primary alkaloid of various species of Cinchona (Rubiaceae). It is also an antimalarial and muscle relaxant (skeletal). Purity 99.45%
Catalog No. 50-287-8932 Supplier TARGETMOL CHEMICALS INC Supplier No. T4333500MG
Also available in 1 g, 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Quinine dihydrochloride (Quinine bimuriate) is a primary alkaloid of various species of Cinchona (Rubiaceae). It is also an antimalarial and muscle relaxant (skeletal). Purity 99.45%