Learn More
Apexbio Technology LLC Quinine HCl Dihydrate 6119-47-7 10g
SDP
Catalog No. 5000090084
Encompass_Preferred
Quinine HCl Dihydrate is a quinoline alkaloid hydrochloride salt presented as a dihydrate form It exerts its biological activity primarily through interference with heme detoxification mechanisms within Plasmodium species disrupting parasite-mediated heme metabolism and inducing cytotoxic free heme accumulation At the molecular level quinoline derivatives may inhibit nucleic acid synthesis affecting DNA replication and transcription events during erythrocytic lifecycle stages Researchers frequently utilize Quinine HCl Dihydrate for studies on antimalarial drug mechanisms parasite resistance pathways and as a reference standard in pharmacological and pharmacokinetic assessments
Catalog No. 50-000-90084 Supplier Apexbio Technology LLC Supplier No. B212310000
Quinine HCl Dihydrate is a quinoline alkaloid hydrochloride salt presented as a dihydrate form It exerts its biological activity primarily through interference with heme detoxification mechanisms within Plasmodium species disrupting parasite-mediated heme metabolism and inducing cytotoxic free heme accumulation At the molecular level quinoline derivatives may inhibit nucleic acid synthesis affecting DNA replication and transcription events during erythrocytic lifecycle stages Researchers frequently utilize Quinine HCl Dihydrate for studies on antimalarial drug mechanisms parasite resistance pathways and as a reference standard in pharmacological and pharmacokinetic assessments