Learn More
Apexbio Technology LLC Quinine HCl Dihydrate 6119-47-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000089394
Encompass_Preferred
Quinine HCl Dihydrate is a quinoline alkaloid hydrochloride salt presented as a dihydrate form It exerts its biological activity primarily through interference with heme detoxification mechanisms within Plasmodium species disrupting parasite-mediated heme metabolism and inducing cytotoxic free heme accumulation At the molecular level quinoline derivatives may inhibit nucleic acid synthesis affecting DNA replication and transcription events during erythrocytic lifecycle stages Researchers frequently utilize Quinine HCl Dihydrate for studies on antimalarial drug mechanisms parasite resistance pathways and as a reference standard in pharmacological and pharmacokinetic assessments
Catalog No. 50-000-89394 Supplier Apexbio Technology LLC Supplier No. B21235.1
Quinine HCl Dihydrate is a quinoline alkaloid hydrochloride salt presented as a dihydrate form It exerts its biological activity primarily through interference with heme detoxification mechanisms within Plasmodium species disrupting parasite-mediated heme metabolism and inducing cytotoxic free heme accumulation At the molecular level quinoline derivatives may inhibit nucleic acid synthesis affecting DNA replication and transcription events during erythrocytic lifecycle stages Researchers frequently utilize Quinine HCl Dihydrate for studies on antimalarial drug mechanisms parasite resistance pathways and as a reference standard in pharmacological and pharmacokinetic assessments